ENA444725665 Enamine

3-(chloromethyl)-1-methanesulfonylpyrrolidine

  • MDL number MFCD18351471
  • Cas Number : 1339708-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202893

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202893
Availability details for
Quantity
  • Availability
  • Add to Cart