ENA444725667 Enamine

2-bromo-4-cyanobenzene-1-sulfonyl chloride

  • MDL number MFCD13634254
  • Cas Number : 1098387-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31013

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31013
Availability details for
Quantity
  • Availability
  • Add to Cart