ENA444725669 Enamine

ethyl (1R,3S,4S)-2-azabicyclo[2.2.1]heptane-3-carboxylate hydrochloride

  • MDL number MFCD08460283
  • Cas Number : 188057-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36532
Availability details for
Quantity
  • Availability
  • Add to Cart