ENA444725670 Enamine

3-(morpholine-4-carbonyl)benzene-1-sulfonyl chloride

  • MDL number MFCD09817292
  • Cas Number : 1016672-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39625

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39625
Availability details for
Quantity
  • Availability
  • Add to Cart