ENA444725676 Enamine

methyl 4-(4-cyanophenyl)-2,4-dioxobutanoate

  • MDL number MFCD10693634
  • Cas Number : 1152566-31-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51417

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51417
Availability details for
Quantity
  • Availability
  • Add to Cart