ENA444725679 Enamine

4-[(2-methylquinolin-4-yl)methoxy]benzoic acid

  • MDL number MFCD26394838
  • Cas Number : 362488-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52242

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52242
Availability details for
Quantity
  • Availability
  • Add to Cart