ENA444725684 Enamine

N-[(2-chlorophenyl)methyl]acetamide

  • MDL number MFCD02335711
  • Cas Number : 57058-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203303

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203303
Availability details for
Quantity
  • Availability
  • Add to Cart