ENA444725685 Enamine

2,2,2-trifluoroethyl N-(2-oxo-2,3-dihydro-1H-1,3-benzodiazol-5-yl)carbamate

  • MDL number MFCD21373484
  • Cas Number : 1482630-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56597

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56597
Availability details for
Quantity
  • Availability
  • Add to Cart