ENA444725688 Enamine

2-(3-iodo-4,5-dimethoxyphenyl)acetonitrile

  • MDL number MFCD20232364
  • Cas Number : 138491-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59110

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59110
Availability details for
Quantity
  • Availability
  • Add to Cart