ENA444725689 Enamine

4-chloro-2-(pyrrolidin-1-yl)pyridine

  • MDL number MFCD14702601
  • Cas Number : 1209459-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202894

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202894
Availability details for
Quantity
  • Availability
  • Add to Cart