ENA444725690 Enamine

1-(5-methyl-1,2-oxazol-4-yl)ethan-1-one

  • MDL number MFCD01935998
  • Cas Number : 6497-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 29-30
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60284

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60284
Availability details for
Quantity
  • Availability
  • Add to Cart