ENA444725691 Enamine

1H-1,2,3-benzotriazole-5-sulfonyl chloride

  • MDL number MFCD12022586
  • Cas Number : 70938-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60331

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60331
Availability details for
Quantity
  • Availability
  • Add to Cart