ENA444725696 Enamine

1-methoxy-4,4-dimethylpentan-3-amine

  • MDL number MFCD16807943
  • Cas Number : 1248952-18-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202766

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202766
Availability details for
Quantity
  • Availability
  • Add to Cart