ENA444725700 Enamine

methyl 3-(chloromethyl)benzoate

  • MDL number MFCD00157184
  • Cas Number : 34040-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64432

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P271,P272,P280,P301+P312,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P333+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-64432
pictograms GHS05,GHS07
hstatements H302,H314,H315,H317,H335
Availability details for
Quantity
  • Availability
  • Add to Cart