ENA444725701 Enamine

2,2-dimethyl-2,3-dihydro-1-benzofuran-7-sulfonyl chloride

  • MDL number MFCD19160572
  • Cas Number : 87254-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64478

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64478
Availability details for
Quantity
  • Availability
  • Add to Cart