ENA444725710 Enamine

5-bromo-2-chloro-4-(pyrrolidin-1-yl)pyrimidine

  • MDL number MFCD09743871
  • Cas Number : 57054-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68535

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68535
Availability details for
Quantity
  • Availability
  • Add to Cart