ENA444725711 Enamine

3-nitro-N-propylaniline

  • MDL number MFCD00051836
  • Cas Number : 87035-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69276

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69276
Availability details for
Quantity
  • Availability
  • Add to Cart