ENA444725713 Enamine

methyl 2-(benzylamino)-2-methylpropanoate hydrochloride

  • MDL number MFCD28348353
  • Cas Number : 1803597-78-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69734

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69734
Availability details for
Quantity
  • Availability
  • Add to Cart