ENA444725728 Enamine

6-(3-methyl-2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanoic acid

  • MDL number MFCD11643026
  • Cas Number : 52898-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202896

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202896
Availability details for
Quantity
  • Availability
  • Add to Cart