ENA444725731 Enamine

4-methoxy-3-methylpyridine

  • MDL number MFCD06253896
  • Cas Number : 96609-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80953

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80953
Availability details for
Quantity
  • Availability
  • Add to Cart