ENA444725768 Enamine

5-chloro-3,4-dimethyl-1,2-oxazole

  • MDL number MFCD18449673
  • Cas Number : 37543-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99080

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99080
Availability details for
Quantity
  • Availability
  • Add to Cart