ENA447955637 Enamine

[cis-4-fluoro-1-[(2-methyl-1,3-thiazol-4-yl)methyl]pyrrolidin-2-yl]methanamine

  • MDL number MFCD28657146
  • Cas Number : 1820569-71-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-159416
Availability details for
Quantity
  • Availability
  • Add to Cart