ENA447955715 Enamine

1-tert-butyl 3-methyl 2-(propan-2-yl)pyrrolidine-1,3-dicarboxylate

  • MDL number MFCD28397365
  • Cas Number : 1803611-69-9
  • Molecular Weight : 271.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-200168

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200168
Availability details for
Quantity
  • Availability
  • Add to Cart