ENA447955726 Enamine

6-methoxynaphthalen-2-amine hydrochloride

  • MDL number MFCD00081153
  • Cas Number : 3991-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-201890

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-201890
Availability details for
Quantity
  • Availability
  • Add to Cart