ENA447955727 Enamine

2,2-dicyclobutylethan-1-ol

  • MDL number MFCD28397374
  • Cas Number : 1803606-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-201903

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-201903
Availability details for
Quantity
  • Availability
  • Add to Cart