ENA447955730 Enamine

tert-butyl 3-[1-(2,2,2-trifluoroethyl)-1H-1,2,4-triazol-3-yl]morpholine-4-carboxylate

  • MDL number MFCD28383754
  • Cas Number : 1803586-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-201924

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-201924
Availability details for
Quantity
  • Availability
  • Add to Cart