ENA447955731 Enamine

tert-butyl 3-(1-benzyl-1H-1,2,4-triazol-5-yl)morpholine-4-carboxylate

  • MDL number MFCD28383755
  • Cas Number : 1803602-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-201925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-201925
Availability details for
Quantity
  • Availability
  • Add to Cart