ENA447955733 Enamine

1-{[4-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}-4-methylpiperazine

  • MDL number MFCD28383757
  • Cas Number : 2246633-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-201952

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-201952
Availability details for
Quantity
  • Availability
  • Add to Cart