ENA447955734 Enamine

2-[4-(4-methylpiperazin-1-yl)phenyl]acetamide

  • MDL number MFCD28383758
  • Cas Number : 1333468-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202001

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202001
Availability details for
Quantity
  • Availability
  • Add to Cart