ENA447955742 Enamine

3,8-diazatricyclo[7.3.1.0,2,7]trideca-2(7),3,5-trien-9-ol

  • MDL number MFCD28383762
  • Cas Number : 1803585-71-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202589

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202589
Availability details for
Quantity
  • Availability
  • Add to Cart