ENA447955754 Enamine

tert-butyl 4-cyano-1H-pyrrolo[3,2-c]pyridine-1-carboxylate

  • MDL number MFCD28397385
  • Cas Number : 1803586-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203331

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203331
Availability details for
Quantity
  • Availability
  • Add to Cart