ENA447955755 Enamine

1-[(tert-butoxy)carbonyl]-1H-pyrrolo[3,2-c]pyridin-5-ium-5-olate

  • MDL number MFCD28397386
  • Cas Number : 1803610-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203332

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203332
Availability details for
Quantity
  • Availability
  • Add to Cart