ENA447955756 Enamine

[5-(dimethoxymethyl)-2-fluorophenyl]methanol

  • MDL number MFCD28397387
  • Cas Number : 334019-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203352
Availability details for
Quantity
  • Availability
  • Add to Cart