ENA447955758 Enamine

2-[(5-bromo-1H-indol-3-yl)methyl]-2-acetamidopropanedioic acid

  • MDL number MFCD28383775
  • Cas Number : 1803601-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203560

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203560
Availability details for
Quantity
  • Availability
  • Add to Cart