ENA447955759 Enamine

1,3-diethyl 2-[(5-bromo-1H-indol-3-yl)methyl]-2-acetamidopropanedioate

  • MDL number MFCD28383776
  • Cas Number : 75816-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203561

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203561
Availability details for
Quantity
  • Availability
  • Add to Cart