ENA447955761 Enamine

ethyl 2-(cyclopropanesulfonyl)-2-methylpropanoate

  • MDL number MFCD28383777
  • Cas Number : 1803604-80-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203711

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203711
Availability details for
Quantity
  • Availability
  • Add to Cart