ENA447955764 Enamine

5-[3-(trifluoromethoxy)phenyl]-1,3,4-thiadiazol-2-amine hydrochloride

  • MDL number MFCD28397393
  • Cas Number : 1803611-19-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203775

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203775
Availability details for
Quantity
  • Availability
  • Add to Cart