ENA447955765 Enamine

tert-butyl N-{2-[5-(4-chlorophenyl)-4H-1,2,4-triazol-3-yl]ethyl}carbamate

  • MDL number MFCD28383780
  • Cas Number : 1803604-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203784

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203784
Availability details for
Quantity
  • Availability
  • Add to Cart