ENA447955767 Enamine

4-(1-methoxy-2-methylpropan-2-yl)piperidine hydrochloride

  • MDL number MFCD28383782
  • Cas Number : 1803561-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203846
Availability details for
Quantity
  • Availability
  • Add to Cart