ENA447955768 Enamine

tert-butyl 4-(3,4-difluorophenyl)-1,4-diazepane-1-carboxylate

  • MDL number MFCD28383783
  • Cas Number : 868064-73-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203864

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203864
Availability details for
Quantity
  • Availability
  • Add to Cart