ENA447955769 Enamine

tert-butyl 4-(3-chloro-2-methylphenyl)-1,4-diazepane-1-carboxylate

  • MDL number MFCD28383784
  • Cas Number : 1803589-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203865

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203865
Availability details for
Quantity
  • Availability
  • Add to Cart