ENA447955770 Enamine

2-[3-(2-methylpropoxy)phenyl]acetonitrile

  • MDL number MFCD28383785
  • Cas Number : 1803603-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203904

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203904
Availability details for
Quantity
  • Availability
  • Add to Cart