ENA447955772 Enamine

ammonium 5-chloro-1,1,3-trioxo-2,3-dihydro-1lambda6,2-benzothiazol-2-ide

  • MDL number MFCD28383787

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data NH4+
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203942

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203942
Availability details for
Quantity
  • Availability
  • Add to Cart