ENA447955774 Enamine

5H,6H,7H-cyclopenta[b]pyridine-7-carboxylic acid hydrochloride

  • MDL number MFCD28383789
  • Cas Number : 1610919-99-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203980

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203980
Availability details for
Quantity
  • Availability
  • Add to Cart