ENA447955775 Enamine

3-amino-6-(trifluoromethyl)-1,2-dihydropyridin-2-one hydrochloride

  • MDL number MFCD28397394
  • Cas Number : 1803587-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204057

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204057
Availability details for
Quantity
  • Availability
  • Add to Cart