ENA447955780 Enamine

1-(oxan-4-yl)-1-[3-(trifluoromethyl)phenyl]methanamine hydrochloride

  • MDL number MFCD28383796
  • Cas Number : 1803591-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204203

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204203
Availability details for
Quantity
  • Availability
  • Add to Cart