ENA447955782 Enamine

2-chloro-5-propylbenzene-1-sulfonyl chloride

  • MDL number MFCD28383797
  • Cas Number : 1803595-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204287

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204287
Availability details for
Quantity
  • Availability
  • Add to Cart