ENA447955783 Enamine

2-chloro-5-(propan-2-yl)benzene-1-sulfonyl chloride

  • MDL number MFCD28383798
  • Cas Number : 1803597-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204289

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204289
Availability details for
Quantity
  • Availability
  • Add to Cart