ENA447955796 Enamine

6-amino-4-(2,2-difluoroethyl)-3,4-dihydro-2H-1,4-benzothiazin-3-one

  • MDL number MFCD28383810
  • Cas Number : 1803561-78-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204483

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204483
Availability details for
Quantity
  • Availability
  • Add to Cart