ENA447955801 Enamine

5-(4-hydroxypiperidin-1-yl)-2-(piperidin-3-yl)-2,3-dihydropyridazin-3-one; carbonic acid

  • MDL number MFCD28383815
  • Cas Number : 1803561-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data OC(=O)O
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204522

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204522
Availability details for
Quantity
  • Availability
  • Add to Cart